Skip to Content
MilliporeSigma
All Photos(1)

Documents

749001

Sigma-Aldrich

4,6-Dioxoheptanoic acid-3,4,5,6,7-13C5

99 atom % 13C, 97% (CP)

Synonym(s):

Succinylacetone-3,4,5,6,7-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CO13CH213CO13CH2CH2COOH
CAS Number:
Molecular Weight:
163.12
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.13

isotopic purity

99 atom % 13C

Quality Level

Assay

97% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

66-67 °C (lit.)

mass shift

M+5

storage temp.

2-8°C

SMILES string

[13CH3][13C](=O)[13CH2][13C](=O)[13CH2]CC(O)=O

InChI

1S/C7H10O4/c1-5(8)4-6(9)2-3-7(10)11/h2-4H2,1H3,(H,10,11)/i1+1,2+1,4+1,5+1,6+1

InChI key

WYEPBHZLDUPIOD-JBSMRZEESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service