Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748854

Sigma-Aldrich

Iodotrimethylsilane solution

1 M in dichloromethane

Synonym(s):

Iodotrimethylsilane solution, TMS iodine, TMS-I, TMSI, Trimethyliodosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9ISi
Molecular Weight:
200.09
UNSPSC Code:
12352101
NACRES:
NA.22

form

liquid

concentration

1 M in dichloromethane

refractive index

n20/D 1.433

density

1.334 g/mL at 25 °C

storage temp.

2-8°C

General description

Iodotrimethylsilane [(CH3)3SiI] is an important and versatile organosilicon reagent with wide applications in organic synthesis. It is used as a neutral nucleophilic reagent, trimethylsilylating agent, Lewis acid catalyst, reducing agent and dehydrating agent in many organic reactions.

Application

Iodotrimethylsilane can be used as:
  • A reagent in the hydroiodation of ynamides to yield the important building blocks (E)-α-iodoenamides.
  • A reagent in the synthesis of (1-iodovinyl) arenes by hydroiodation of trimethylsilyl ethynylarenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-23.8 °F

Flash Point(C)

-31 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step synthesis of (1-iodovinyl) arenes from trimethylsilyl ethynylarene through iodotrimethylsilane-mediated hydroiodation
Sato AH, et al.
Tetrahedron Letters, 53(28), 3585-3589 (2012)
Iodotrimethylsilane
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2005)
Regio-and stereospecific synthesis of (E)-α-iodoenamide moieties from ynamides through iodotrimethylsilane-mediated hydroiodation
Sato AH, et al.
Tetrahedron Letters, 54(10), 1309-1311 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service