Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

747688

Sigma-Aldrich

Copper(II) trifluoromethanesulfonimide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4CuF12N2O8S4 · xH2O
CAS Number:
Molecular Weight:
623.84 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: copper
reagent type: catalyst

mp

159-165 °C

storage temp.

2-8°C

SMILES string

O=S(N([Cu]N(S(=O)(C(F)(F)F)=O)S(=O)(C(F)(F)F)=O)S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.O

InChI

1S/2C2F6NO4S2.Cu.H2O/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;;/h;;;1H2/q2*-1;+2;

InChI key

SOSOQNLWNNNBFY-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service