Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

739731

Sigma-Aldrich

2-Aminooxazole

97%

Synonym(s):

2-Oxazolamine, Oxazole-2-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2O
CAS Number:
Molecular Weight:
84.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

90-95 °C

storage temp.

2-8°C

SMILES string

Nc1ncco1

InChI

1S/C3H4N2O/c4-3-5-1-2-6-3/h1-2H,(H2,4,5)

InChI key

ACTKAGSPIFDCMF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bini Mathew et al.
European journal of medicinal chemistry, 210, 112952-112952 (2020-11-04)
ALS is a rare type of progressive neurological disease with unknown etiology. It results in the gradual degeneration and death of motor neurons responsible for controlling the voluntary muscles. Identification of mutations in the superoxide dismutase (SOD) 1 gene has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service