Skip to Content
MilliporeSigma
All Photos(1)

Documents

739286

Sigma-Aldrich

1-Hexanethiol

99% (GC)

Synonym(s):

1-Mercaptohexane, Hexyl mercaptan, MCH, Mercaptan C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SH
CAS Number:
Molecular Weight:
118.24
Beilstein:
1731295
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% (GC)

form

liquid

refractive index

n20/D 1.450

bp

150-154 °C (lit.)

mp

−81-−80 °C (lit.)

density

0.832 g/mL at 25 °C

SMILES string

CCCCCCS

InChI

1S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

PMBXCGGQNSVESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanethiol (HT) is a non-polar alkanethiol that forms a self-assembled monolayer (SAM) on a variety of surfaces. It facilitates the immobilization of surface atoms and can be used in chemical sensing processes.

Application

HT forms a SAM on gold surfaces such as electrodes, nanoparticles, and films for a wide range of applications which include chemiresistor sensors and electrochemical detection.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.0 °F

Flash Point(C)

38.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensing characteristics of NIR localized surface plasmon resonances in gold nanorings for application as ultrasensitive biosensors.
Larsson EM, et al.
Nano Letters, 7(5), 1256-1263 (2007)
XPS investigations on the interactions of 1, 6-hexanedithiol/Au (1 1 1) layers with metallic and ionic silver species.
Esplandiu MJ and Noeske P-LM
Applied Surface Science, 199(1-4), 166-182 (2002)
Detection of organics in aqueous solution using gold nanoparticles modified with mixed monolayers of 1-hexanethiol and 4-mercaptophenol.
Chow E, et al.
Sensors and Actuators B, Chemical, 143(2), 704-711 (2010)
Ultrasensitive detection of toxic cations through changes in the tunnelling current across films of striped nanoparticles.
Cho ES, et al.
Testing, 11(11), 978-978 (2012)
Effect of Composition on the Catalytic Properties of Mixed-Ligand-Coated Gold Nanoparticles.
Ghosh A, et al.
Angewandte Chemie (International Edition in English), 50(34), 7900-7905 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service