Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

737267

Sigma-Aldrich

Di(1-adamantyl)chlorophosphine

97%

Synonym(s):

Bis(1-adamantyl)phosphinous chloride, Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphinous chloride, Diadamantylchlorophosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30ClP
CAS Number:
Molecular Weight:
336.88
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

impurities

may contain ≤1% CCl4

mp

168-173 °C

functional group

phosphine

SMILES string

ClP([C@]12C[C@H]3C[C@H](C[C@H](C3)C1)C2)[C@]45C[C@H]6C[C@H](C[C@H](C6)C4)C5

InChI

1S/C20H30ClP/c21-22(19-7-13-1-14(8-19)3-15(2-13)9-19)20-10-16-4-17(11-20)6-18(5-16)12-20/h13-18H,1-12H2/t13-,14+,15-,16-,17+,18-,19-,20-

InChI key

BVOJCPQWSXCCKU-RWTPHSKKSA-N

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Ozone 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation - Water-react 3

Target Organs

Liver,Kidney

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service