Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735345

Sigma-Aldrich

5-Carboxythiophene-2-boronic acid pinacol ester

96%

Synonym(s):

5-Carboxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, 5-Carboxy-2-thienylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BO4S
CAS Number:
Molecular Weight:
254.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

176-181 °C

functional group

carboxylic acid

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(s2)C(O)=O

InChI

1S/C11H15BO4S/c1-10(2)11(3,4)16-12(15-10)8-6-5-7(17-8)9(13)14/h5-6H,1-4H3,(H,13,14)

InChI key

YLZRZHOUXZZBTF-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service