Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735280

Sigma-Aldrich

3-Fluoro-4-(hydroxymethyl)benzonitrile

97%

Synonym(s):

4-Cyano-2-fluorobenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FNO
CAS Number:
Molecular Weight:
151.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

63-68 °C

density

1.27 g/mL at 25 °C

functional group

fluoro
hydroxyl
nitrile

SMILES string

OCc1ccc(cc1F)C#N

InChI

1S/C8H6FNO/c9-8-3-6(4-10)1-2-7(8)5-11/h1-3,11H,5H2

InChI key

YGAURRAHPYQHDC-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service