Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733873

Sigma-Aldrich

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone

≥98 atom %, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C5D3H11N2O4S
CAS Number:
Molecular Weight:
227.27
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom %

Quality Level

Assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

132-135 °C (lit.)

mass shift

M+5

storage temp.

−20°C

SMILES string

[2H][13C]([2H])([2H])N[13C](=O)O\N=C\C(C)(C)S(C)(=O)=O

InChI

1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10)/b9-5+/i3+1D3,6+1

InChI key

YRRKLBAKDXSTNC-FONWFMSDSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service