Skip to Content
MilliporeSigma
All Photos(1)

Documents

732338

Sigma-Aldrich

Hexanal-d12

≥98 atom % D, 96% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)4CDO
CAS Number:
Molecular Weight:
112.23
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Assay

96% (CP)

form

liquid

bp

130-131 °C

density

0.91 g/mL at 25 °C (lit.)

mass shift

M+12

storage temp.

2-8°C

SMILES string

[2H]C(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D

InChI key

JARKCYVAAOWBJS-PSDCQUMKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Rohfritsch et al.
Journal of agricultural and food chemistry, 67(12), 3511-3520 (2019-03-01)
Long-chain polyunsaturated fatty acids are highly susceptible to lipid oxidation which causes undesirable odors and flavors in food. We present the development, validation, and application of a semiquantitative screening method to monitor volatile and nonvolatile carbonyl compounds generated from lipids
Xinyi Zhang et al.
Journal of agricultural and food chemistry, 67(12), 3502-3510 (2019-02-28)
This work outlines a rapid novel methodology for the direct quantitation of 12 volatile aldehyde compounds related to oxidative off-flavors in wine, by measuring the combined free and hydrogen-sulfite-bound forms of each aldehyde compound, consisting of four general aldehydes, four

Articles

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service