Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732214

Sigma-Aldrich

4-Bromo-3-thiophenecarboxylic acid

97%

Synonym(s):

4-Bromothiophene-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrO2S
CAS Number:
Molecular Weight:
207.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

158-163 °C

functional group

bromo
carboxylic acid

SMILES string

OC(=O)c1cscc1Br

InChI

1S/C5H3BrO2S/c6-4-2-9-1-3(4)5(7)8/h1-2H,(H,7,8)

InChI key

APBIZVFQQBCAAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon C Fillmore et al.
ACS omega, 5(36), 22914-22925 (2020-09-22)
A pathway to a range of diverse heterocycles was developed using a nucleophilic cyclization strategy. Lactams and ene-imines are accessed in a few steps from a common precursor, and these moieties are further elaborated to directly provide pyrroles or pyridines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service