Skip to Content
MilliporeSigma
All Photos(1)

Documents

731641

Sigma-Aldrich

16-α-Hydroxyestrone-2,3,4-13C3

99 atom % 13C, 98% (CP)

Synonym(s):

1,3,5(10)-Estratriene-3,16α-diol-17-one-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C15H22O3
CAS Number:
Molecular Weight:
289.34
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4[13cH][13c](O)[13cH]cc34)[C@@H]1C[C@@H](O)C2=O

InChI

1S/C18H22O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-16,19-20H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,18+/m1/s1/i3+1,8+1,11+1

InChI key

WPOCIZJTELRQMF-SUZPQBOQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

To best serve customer requests, each order is packaged and shipped on demand approximately 2 weeks after receipt of an order. If you have questions regarding availability and price, please contact the Stable Isotope Customer Service Group at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at isosales@sial.com.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service