Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

730068

Sigma-Aldrich

rac-Glyceryl-d5-2-linoleate-3-oleate-1-palmitate

98 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55D5H95O6
Molecular Weight:
862.41
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

95% (CP)

form

liquid

mass shift

M+5

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])(OC(=O)CCCCCCCCCCCCCCC)C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C([2H])([2H])OC(=O)CCCCCCCC\C=C/CCCCCCC

InChI

1S/C55H100O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h17,20,22,25-26,28,52H,4-16,18-19,21,23-24,27,29-51H2,1-3H3/b20-17-,25-22-,28-26-/i50D2,51D2,52D

InChI key

FHSLPRLHVWRPNQ-PBYMJFEOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service