Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

729833

Sigma-Aldrich

2-N-Morpholinoethyl methacrylate

contains MEHQ as inhibitor, 95%

Synonym(s):

2-Methylprop-2-enoate, 2-Morpholin-4-ylethyl, MEMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

contains

MEHQ as inhibitor

SMILES string

CC(=C)C(=O)OCCN1CCOCC1

InChI

1S/C10H17NO3/c1-9(2)10(12)14-8-5-11-3-6-13-7-4-11/h1,3-8H2,2H3

InChI key

MNZNJOQNLFEAKG-UHFFFAOYSA-N

General description

2-N-Morpholinoethyl methacrylate (MEMA) is a methacrylate monomer featuring a morpholino group that enhances its reactivity and solubility in various solvents. Its hydrophilicity, attributed to the tertiary amine in the morpholino structure, makes MEMA suitable for numerous applications in polymer chemistry. It can undergo various polymerization techniques, including radical and anionic polymerization. MEMA is particularly valuable in biomedical applications, including drug delivery systems, medical adhesives, dental composites, tissue engineering, and controlled drug release. It is utilized in the development of biocompatible materials, polymer matrices, hydrogels, and scaffolds. Beyond biomedical uses, MEMA finds applications in coatings, electronics, optical materials, and the textile industry.

Application

2-N-Morpholinoethyl methacrylate can be used as:
  • A monomer for synthesizing pH-responsive amphiphilic polymers. These polymers are particularly useful in controlled release drug delivery systems due to their ability to respond to changes in pH, which is a critical factor in various physiological environments.
  • A crosslinking agent during the synthesis of the hydrogel nanocomposites. This crosslinking is vital for improving the mechanical stability and structural integrity of the hydrogels.
  • A monomer for the synthesis of thermoresponsive polymers, which are used for the applications in smart coatings and responsive drug delivery systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucilla Iacumin et al.
Microorganisms, 8(10) (2020-10-25)
Recently, specific dry-cured hams have started to be produced in San Daniele and Parma areas. The ingredients are similar to protected denomination of origin (PDO) produced in San Daniele or Parma areas, and include pork leg, coming from pigs bred
Lucilla Iacumin et al.
Food microbiology, 62, 188-195 (2016-11-28)
Speck is a meat product obtained from the deboned leg of pork that is salted, smoked and seasoned for four to six months. During speck seasoning, Eurotium rubrum and Penicillium solitum grow on the surface and collaborate with other moulds
Priyanshi Agnihotri et al.
Soft matter, 16(33), 7845-7859 (2020-08-07)
Herein, we report synthesis of thermoresponsive poly(N-isopropylaccrylamide) (PNIPAM) microgels with short oligo(ethylene glycol) (OEG) chain comonomers (1 to 4/5 repeating unit) by surfactant-free precipitation copolymerization. The efficient incorporation of the comonomers was confirmed by a complete set of characterization methods
Ujjwal Kumar Chowdhury et al.
Texas Heart Institute journal, 47(2), 86-95 (2020-07-01)
We evaluated aortic tissue specimens from patients undergoing tetralogy of Fallot repair, to determine whether histologic abnormalities affect postsurgical aortic remodeling and other patient-related variables. Using light microscopy, we studied full-thickness aortic wall tissue operatively excised from 118 consecutive patients
Linyong Song et al.
Journal of dentistry, 43(9), 1132-1139 (2015-07-07)
The objective of this work was to investigate the polymerization behavior, neutralization capability, and mechanical properties of dentin adhesive formulations with the addition of the tertiary amine co-monomer, 2-N-morpholinoethyl methacrylate (MEMA). A co-monomer mixture based on HEMA/BisGMA (45/55, w/w) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service