727725
1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate
95%
Synonym(s):
BMPyrrOTf
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H20F3NO3S
CAS Number:
Molecular Weight:
291.33
Beilstein:
9820503
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0% (T)
95%
form
liquid
impurities
≤0.3% water
functional group
fluoro
triflate
SMILES string
[O-]S(=O)(=O)C(F)(F)F.CCCC[N+]1(C)CCCC1
InChI
1S/C9H20N.CHF3O3S/c1-3-4-7-10(2)8-5-6-9-10;2-1(3,4)8(5,6)7/h3-9H2,1-2H3;(H,5,6,7)/q+1;/p-1
InChI key
WZJDNKTZWIOOJE-UHFFFAOYSA-M
Application
1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate ([BMPy][OTf]) is an ionic liquid that can be used as a solvent in:
([BMPy][OTf]) can also be used as an electrolyte in supercapacitor applications.
- Rhodium-catalyzed regioselective hydroformylation reactions.
- Direct asymmetric aldol condensation reaction.
- Desulfurization of fuels.
- Nucleophilic aromatic substitution reactions.
([BMPy][OTf]) can also be used as an electrolyte in supercapacitor applications.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
An improved protocol for the direct asymmetric aldol reaction in ionic liquids, catalysed by onium ion-tagged prolines
Lombardo M, et al.
Advanced Synthesis & Catalysis, 349(11-12), 2061-2065 (2007)
Andrzej Lewandowski et al.
Journal of solution chemistry, 42(2), 251-262 (2013-03-14)
The electrochemical behavior of cobaltocenium has been studied in a number of room temperature aprotic ionic liquids. Well defined, diffusion controlled, anodic and cathodic peaks were found for the Cc
Effect of the cation and anion of the ionic liquid on desulfurization of model fuels
Domanska U and Wlazlo M
Fuel: The Science and Technology of Fuel and Energy, 134(11-12), 114-125 (2014)
An improved protocol for the direct asymmetric aldol reaction in ionic liquids, catalysed by onium ion-tagged prolines
Lombardo M, et al.
advanced synthesis and catalysis, 349(11-12), 2061-2065 (2007)
Ionic liquids as designer solvents for nucleophilic aromatic substitutions
Newington I, et al.
Organic Letters, 9(25), 5247-5250 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service