Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725641

L-Arginine-1-13C dihydrate

≥99 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H14N4O2 · 2H2O
Molecular Weight:
211.22
UNSPSC Code:
12352209

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

form

powder

mass shift

M+1

SMILES string

NC(NCCC[C@H](N)[13C](O)=O)=N.O.O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service