Skip to Content
MilliporeSigma
All Photos(1)

Documents

721824

Sigma-Aldrich

1-Boc-2-(aminomethyl)pyrrolidine

Synonym(s):

tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

refractive index

n20/D 1.472
n20/D 1.472

density

1.014 g/mL at 25 °C
1.014 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCCC1CN

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-6-4-5-8(12)7-11/h8H,4-7,11H2,1-3H3

InChI key

SOGXYCNKQQJEED-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service