Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72060

Sigma-Aldrich

Sodium metavanadate

≥98.0% (RT)

Synonym(s):

Sodium (meta)vanadate, Sodium trioxovanadate, Sodium vanadate(V), Sodium vanadium oxide, Sodium vanadium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaVO3
CAS Number:
Molecular Weight:
121.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

≥98.0% (RT)

form

powder

reaction suitability

reagent type: catalyst
core: vanadium

technique(s)

cell culture | mammalian: suitable

SMILES string

[Na+].[O-][V](=O)=O

InChI

1S/Na.3O.V/q+1;;;-1;

InChI key

CMZUMMUJMWNLFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium metavanadate (SMV) is a vanadium salt that can be used as a corrosion inhibitor with good inhibition efficiency (98%) at 200 ppm. It can be used in the protection of carbon steel.

Application


  • Study of electrical properties of cobalt oxide-doped sodium meta vanadate at different frequencies: This study explores the electrical properties of sodium metavanadate doped with cobalt oxide, emphasizing its significance in electrical insulation applications (Saleh & Deeb, 2022).

  • Chemical synthesis of vanadium oxide (V2O5) nanoparticles prepared by sodium metavanadate: This research demonstrates a simple chemical method to synthesize vanadium oxide nanoparticles using sodium metavanadate, highlighting potential uses in nanotechnology and material sciences (Farahmandjou & Abaeiyan, 2017).

  • Oxidative dehydrogenation of propane on silica-supported vanadyl sites promoted with sodium metavanadate: Discusses the catalytic properties of sodium metavanadate in the oxidative dehydrogenation of propane, underlining its potential in industrial chemical processes (Nadjafi et al., 2020).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zsolt Kása et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-24)
In the present study, additive-free, pH-driven, hydrothermal crystallization was used to obtain shape-tailored monoclinic BiVO4 photocatalysts. The as-prepared BiVO4 products were systematically characterized, uncovering their crystallographic, morphologic and optical properties, while their applicability was verified in the visible light-driven photodegradation
Corrosion inhibition by green inhibitor: sodium metavanadate-spirulina system
Sribhurathy V and Rejendran S
Chemical Science Review and Letters, 1(1), 25-29 (2012)
Corrosion inhibition of carbon steel by sodium metavanadate
Sribharathy VG and Rajendran S
Journal of Electrochemical Science and Engineering, 2(3), 121-131 (2012)
Gwendolyn Moise et al.
Biochemistry, 54(42), 6490-6500 (2015-10-08)
Catalysis in protein tyrosine phosphatases (PTPs) involves movement of a protein loop called the WPD loop that brings a conserved aspartic acid into the active site to function as a general acid. Mutation of the tryptophan in the WPD loop
Oluwaseun Mustapha et al.
Environmental toxicology and pharmacology, 38(1), 98-111 (2014-06-14)
Vanadium exposure has been known to lead to lipid peroxidation, demyelination and oligodendrocytes depletion. We investigated behaviour and glial reactions in juvenile mice after early neonatal exposure to vanadium, and examined the direct effects of vanadium in oligodendrocyte progenitor cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service