Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

719552

Sigma-Aldrich

17β-Estradiol-2,3,4-13C3

99 atom % 13C, 98% (CP)

Synonym(s):

1,3,5-Estratriene-3-17β-diol-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C15H24O2
CAS Number:
Molecular Weight:
275.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

178-179 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

O[13C]1=[13CH]C=C(C2=[13CH]1)[C@]3([H])[C@](CC2)([H])[C@@](CC[C@@H]4O)([H])[C@]4(C)CC3

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3+1,10+1,12+1

InChI key

VOXZDWNPVJITMN-CLMZCXLCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service