Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

716812

Sigma-Aldrich

Gold nanorods

10 nm diameter, λmax, 780 nm, dispersion in H2O

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

colloidal suspension
dispersion in H2O
nanorod

contains

CTAB as stabilizer

concentration

>30 μg/mL in H2O

diam. × L

10 nm × 38 nm , ±10%

impurities

<0.1% CTAB
<0.1% ascorbic acid

diameter

10 nm

pH

3-5

density

1.00 g/mL at 25 °C

λmax

780 nm

Mw/Mn

(<10% CV, monodispersity)
>95 (rods)

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Zinc standard for biochemical research: A study in Micromachines reported the development of a novel thin-layer flow cell sensor system that utilizes a zinc standard solution for heavy metal ion detection. This application highlights the utility of high-purity zinc standards in enhancing the sensitivity and specificity of biochemical assays, particularly in environmental monitoring and public health sectors (Xiao et al., 2024).


Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service