Skip to Content
MilliporeSigma
All Photos(1)

Documents

716391

Sigma-Aldrich

3-Hydroxy-4-methoxyphenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

127-132 °C

SMILES string

COc1ccc(CC(O)=O)cc1O

InChI

1S/C9H10O4/c1-13-8-3-2-6(4-7(8)10)5-9(11)12/h2-4,10H,5H2,1H3,(H,11,12)

InChI key

BWXLCOBSWMQCGP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Sim et al.
Clinical and experimental hypertension. Part A, Theory and practice, 12(3), 343-353 (1990-01-01)
The levels of dopamine and its metabolites in the adenohypophysis of the normo- and hypertensive rats were determined. Dopamine was present as the free and sulfate-conjugated form in the adenohypophysis of the spontaneously hypertensive rat (SHR), Wistar Kyoto (WKY) and
F A Muskiet et al.
Clinical chemistry, 25(10), 1708-1713 (1979-10-01)
We describe the determination of the 4-O-methylated catecholamine metabolites 4-methoxy-3-hydroxyphenylacetic acid (iso-HVA), 4-methoxy-3-hydroxyphenylmandelic acid (iso-VMA) and 4-methoxy-3-hydroxyphenylethylene glycol (iso-MOPEG) in urine with use of mass fragmentography and deuterated internal standards. Normal values, expressed in terms of creatinine, are given as
T Oishi et al.
Journal of neural transmission, 53(2-3), 101-108 (1982-01-01)
The injection of reserpine [2.5 mg/kg, intraperitoneally (i.p.)] increased brain concentration of tyrosine and of the major catecholamine metabolites, 3-methoxy-4-hydroxy-phenylethylene--glycol-sulfate (MHPG-SO4) in the rat. In reserpine-treated animals, tyrosine administration (200 mg/kg, i.p.) caused further increases in brain tyrosine, DOPAC, and
J Chauhan et al.
Journal of chromatography, 183(4), 391-401 (1980-10-10)
The determination of urinary homovanillic, isohomovanillic and vanillylmandelic acids as their trifluoroacetylhexafluoroisopropyl ester derivatives by glass capillary gas-liquid chromatography has been studied. It was shown that even with high column efficiencies a single peak-single compound relationship could not be assumed
Jaroslav Havlik et al.
Nutrients, 12(6) (2020-06-03)
Polyphenols are often ingested alongside dietary fibres. They are both catabolised by, and may influence, the intestinal microbiota; yet, interactions between them and the impact on their resultant microbial products are poorly understood. Dietary fibres (inulin, pectin, psyllium, pyrodextrin, wheat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service