Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

711101

Sigma-Aldrich

Potassium pyridine-3-trifluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BF3KN
CAS Number:
Molecular Weight:
185.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

228-232 °C

SMILES string

[K+].F[B-](F)(F)c1cccnc1

InChI

1S/C5H4BF3N.K/c7-6(8,9)5-2-1-3-10-4-5;/h1-4H;/q-1;+1

InChI key

MNCKCIOWIHHCSC-UHFFFAOYSA-N

Application

An organotrifluoroborate involved in:
  • Nickel-catalyzed cross-coupling reactions and C-O activation
  • Suzuki cross-coupling
  • Copper-catalyzed cross-coupling reactions
  • Stereoselective Mukaiyama aldol reactions

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service