Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

709379

Sigma-Aldrich

Indazole-4-boronic acid hydrochloride

Synonym(s):

Indazol-4-ylboronic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BClN2O2
CAS Number:
Molecular Weight:
198.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

173-177 °C

storage temp.

2-8°C

SMILES string

Cl.OB(O)c1cccc2[nH]ncc12

InChI

1S/C7H7BN2O2.ClH/c11-8(12)6-2-1-3-7-5(6)4-9-10-7;/h1-4,11-12H,(H,9,10);1H

InChI key

PAILGUQNESDYLX-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Application

Reactant for:
  • Preparation of trisubstituted pyrimidines as phosphatidylinositol 3-kinase inhibitors
  • Preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service