Skip to Content
MilliporeSigma
All Photos(1)

Documents

708062

Sigma-Aldrich

Eicosanoic acid-20,20,20-d3

99 atom % D, 98% (CP)

Synonym(s):

Arachic acid-20,20,20-d3, Arachidic acid-20,20,20-d3, Arachidinic acid-20,20,20-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)18COOH
Molecular Weight:
315.55
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

98% (CP)

form

powder

mp

74-76 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)/i1D3

InChI key

VKOBVWXKNCXXDE-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service