Skip to Content
MilliporeSigma

707643

Benzothiazole-2-carboxaldehyde

greener alternative

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H5NOS
CAS Number:
Molecular Weight:
163.20
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

solid

greener alternative product score

old score: 13
new score: 6
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

72-76 °C

functional group

aldehyde

greener alternative category

SMILES string

O=Cc1nc2ccccc2s1

InChI

1S/C8H5NOS/c10-5-8-9-6-3-1-2-4-7(6)11-8/h1-5H

InChI key

RHKPJTFLRQNNGJ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy”, “Less Hazardous Chemical Syntheses”, “Safer Solvents and Auxiliaries” and “Use of Renewable Feedstock”. Click here to view its DOZN scorecard.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library