Skip to Content
MilliporeSigma
All Photos(1)

Documents

703168

Sigma-Aldrich

cis-Bis(isothiocyanato)(2,2′-bipyridyl-4,4′-dicarboxylato)(4,4′-di-nonyl-2′-bipyridyl)ruthenium(II)

95% (NMR)

Synonym(s):

Greatcell Solar®, Z-907 dye

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H52N6O4RuS2
CAS Number:
Molecular Weight:
870.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (NMR)

form

powder

composition

Dye content, ≥90%

mp

196 °C

λmax

531, 314, 295 nm (lit.)

SMILES string

S=C=N[Ru]N=C=S.OC(=O)c1ccnc(c1)-c2cc(ccn2)C(O)=O.CCCCCCCCCc3ccnc(c3)-c4cc(CCCCCCCCC)ccn4

InChI

1S/C28H44N2.C12H8N2O4.2CNS.Ru/c1-3-5-7-9-11-13-15-17-25-19-21-29-27(23-25)28-24-26(20-22-30-28)18-16-14-12-10-8-6-4-2;15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10;2*2-1-3;/h19-24H,3-18H2,1-2H3;1-6H,(H,15,16)(H,17,18);;;/q;;2*-1;+2

InChI key

LALSZYPVVQFXIC-UHFFFAOYSA-N

Application

High-performance ruthenium-based dye used in dye-sensitized solar cells (DSCs) for the sensitization of Titania. Hydrophobic character due to the presence of C9 alkyl chains.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service