Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

702803

Sigma-Aldrich

2,1,3-Benzothiadiazole-4,7-bis(boronic acid pinacol ester)

95%

Synonym(s):

Benzo[c]-1,2,3-thiadiazol-4,7-diyl-4,7-diboronic acid dipinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26B2N2O4S
CAS Number:
Molecular Weight:
388.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

mp

207-212 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(B3OC(C)(C)C(C)(C)O3)c4nsnc24

InChI

1S/C18H26B2N2O4S/c1-15(2)16(3,4)24-19(23-15)11-9-10-12(14-13(11)21-27-22-14)20-25-17(5,6)18(7,8)26-20/h9-10H,1-8H3

InChI key

DHWADSHFLSDMBJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service