Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694967

Sigma-Aldrich

N-(2-Aminoethyl)-4-(trifluoromethyl)benzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4SO2NHCH2CH2NH2
CAS Number:
Molecular Weight:
268.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

111-115 °C

functional group

amine
fluoro

SMILES string

NCCNS(=O)(=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C9H11F3N2O2S/c10-9(11,12)7-1-3-8(4-2-7)17(15,16)14-6-5-13/h1-4,14H,5-6,13H2

InChI key

HOVKSPHQQLXZON-UHFFFAOYSA-N

Application

N-(2-Aminoethyl)-4-(trifluoromethyl)benzenesulfonamide can be used as a ligand to synthesize half-sandwich Rh(III) complex, which is employed as a catalyst for the reduction of NAD+ to NADH by transfer hydrogenation using sodium formate as a hydride source. It is also used to form pentamethylcyclopentadienyl iridium (Cp*Ir)-diamine catalyst for selective transfer hydrogenation of various quinoxalines to synthesize corresponding tetrahydroquinxalines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

pH-regulated transfer hydrogenation of quinoxalines with a Cp* Ir-diamine catalyst in aqueous media
Tan J, et al.
Tetrahedron, 67(34), 6206-6213 (2011)
Joan J Soldevila-Barreda et al.
Journal of inorganic biochemistry, 153, 322-333 (2015-11-26)
Organometallic complexes have the potential to behave as catalytic drugs. We investigate here Rh(III) complexes of general formula [(Cp(x))Rh(N,N')(Cl)], where N,N' is ethylenediamine (en), 2,2'-bipyridine (bpy), 1,10-phenanthroline (phen) or N-(2-aminoethyl)-4-(trifluoromethyl)benzenesulfonamide (TfEn), and Cp(x) is pentamethylcyclopentadienyl (Cp*), 1-phenyl-2,3,4,5-tetramethylcyclopentadienyl (Cp(xPh)) or 1-biphenyl-2,3,4,5-tetramethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service