Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694533

Sigma-Aldrich

2-Fluorocyclohexanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9FO
CAS Number:
Molecular Weight:
116.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.040

bp

111-110 °C/70 mmHg

density

1.040 g/mL at 25 °C

storage temp.

−20°C

SMILES string

FC1CCCCC1=O

InChI

1S/C6H9FO/c7-5-3-1-2-4-6(5)8/h5H,1-4H2

InChI key

VQYOFTVCYSPHPG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.0 °F

Flash Point(C)

71.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service