Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693057

Sigma-Aldrich

(S)-BINAP

Synonym(s):

(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, (S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl, (S)-(−)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (S)-(−)-BINAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67
Beilstein:
5321443
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

crystals

Quality Level

optical activity

[α]20/D -222, c = 0.5 in benzene

mp

238-240 °C (lit.)

functional group

phosphine

SMILES string

P(c8ccccc8)(c7ccccc7)c1c(c6c(cc1)cccc6)c2c3c(ccc2P(c5ccccc5)c4ccccc4)cccc3

InChI

1S/C44H32P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32H

InChI key

MUALRAIOVNYAIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Enantioselective and diastereoselective unpoled carbonyl allylation
  • Syntehsis of organophophine oxides as anittumor agents
  • SN2 halogenation of hydroxy groups
  • Synthesis of BINAP complexes
  • Studies of conformational flexibility of BINAP chelates

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim-Hung Lam et al.
European journal of medicinal chemistry, 45(11), 5527-5530 (2010-09-14)
Following our previously reported pyridinyl phosphine oxides as antitumor agents, we targeted the commercially available C(2)-axial chiral organophosphine ligand catalysts, such as 2,2'-bis(diphenylphosphino)-1,1'-binaphthyl (BINAP) 1 and 2,2',6,6'-tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine (P-Phos) 2 as a convenient source for producing organophosphine oxides as antitumor leads.

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service