Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69230

Sigma-Aldrich

1-Methyl-3-octylimidazolium hexafluorophosphate

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23F6N2P
CAS Number:
Molecular Weight:
340.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

liquid

impurities

≤1.0% water

SMILES string

F[P-](F)(F)(F)(F)F.CCCCCCCC[n+]1ccn(C)c1

InChI

1S/C12H23N2.F6P/c1-3-4-5-6-7-8-9-14-11-10-13(2)12-14;1-7(2,3,4,5)6/h10-12H,3-9H2,1-2H3;/q+1;-1

InChI key

GRCIJNHHTXBJAK-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ainhoa Rubio-Clemente et al.
Journal of environmental sciences (China), 92, 79-94 (2020-05-21)
The photodegradation of anthracene (AN) and benzo[a]pyrene (BaP), two priority polycyclic aromatic hydrocarbons (PAHs), was examined at ultra-trace levels in surface water to elucidate their behaviour under several irradiance values and types of radiation. The emitting flux and the spectrum
Naiwei Gao et al.
Nature communications, 10(1), 547-547 (2019-02-03)
Liquid sensors composed of ionic liquids are rising as alternatives to solid semiconductors for flexible and self-healing electronics. However, the fluidic nature may give rise to leakage problems in cases of accidental damages. Here, we proposed a liquid sensor based
Susan Sadeghi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 201, 134-142 (2018-05-11)
In this study, dispersive liquid-liquid micro-extraction using ionic liquid (IL-DLLME) combined with zero crossing first derivative spectrophotometric method was applied to quantitative determination of triphenylmethane dyes in binary mixtures. The 1-methyl-3-octylimidazolium hexafluorophosphate [OMIM][PF6] ionic liquid was used to extract Brilliant
Sara Jamshidi et al.
Journal of separation science, 42(3), 698-705 (2018-11-19)
In this work, a novel, efficient, and green sorbent, SiO2 @Fe3 O4 has been created and functionalized with 1-butyl-3-methylimidazolium hexafluorophosphate as an ionic liquid. This sorbent was applied for microextraction of four beta blockers, propranolol, metoprolol, atenolol, and alprenolol with
Mohammad Saremi et al.
Mikrochimica acta, 186(4), 254-254 (2019-03-25)
An ultrasensitive electrochemiluminescence (ECL) disposable aptamer sensor (aptasensor) is presented for detection of myocardial infarction biomarker by quantification of troponin I in blood serum. A screen-printed electrode was modified with (a) aptamer-modified gold nanoparticles, (b) cyclometallated iridium(III)-poly-4-vinylpyridine nanoparticles, and (c)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service