Skip to Content
MilliporeSigma
All Photos(1)

Documents

692182

Sigma-Aldrich

(R)-Ru(OAc)2(DM-SEGPHOS®)

Synonym(s):

Diacetato[(R)-5,5′-bis[di(3,5-xylyl)phosphino]-4,4′-bi-1,3-benzodioxole]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H50O8P2Ru
Molecular Weight:
941.94
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

CC(=O)O[Ru]OC(C)=O.Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3ccc4OCOc4c3-c5c6OCOc6ccc5P(c7cc(C)cc(C)c7)c8cc(C)cc(C)c8

InChI

1S/C46H44O4P2.2C2H4O2.Ru/c1-27-13-28(2)18-35(17-27)51(36-19-29(3)14-30(4)20-36)41-11-9-39-45(49-25-47-39)43(41)44-42(12-10-40-46(44)50-26-48-40)52(37-21-31(5)15-32(6)22-37)38-23-33(7)16-34(8)24-38;2*1-2(3)4;/h9-24H,25-26H2,1-8H3;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

ACKYBTAGKAINOG-UHFFFAOYSA-L

Application

Catalyst for:
  • Stereoselective preparation of β-amino amides via ruthenium segphos complex-catalyzed asymmetric reductive amination of β-keto amides

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service