Skip to Content
MilliporeSigma
All Photos(1)

Documents

690643

Sigma-Aldrich

3-Bromo-1-butyne

technical, ≥90% (GC)

Synonym(s):

1-Methylpropargyl bromide, 2-Bromo-3-butyne, 3-Bromobutyne, 3-Butyn-2-yl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC(CH3)CC≡H
CAS Number:
Molecular Weight:
132.99
Beilstein:
1098353
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.472

storage temp.

2-8°C

SMILES string

CC(Br)C#C

InChI

1S/C4H5Br/c1-3-4(2)5/h1,4H,2H3

InChI key

PYJVGTWBTIEAMV-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

76.6 °F

Flash Point(C)

24.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service