Skip to Content
MilliporeSigma
All Photos(3)

Documents

688746

Sigma-Aldrich

(S,S)-2,2′-Bipyrrolidine D-tartrate trihydrate

99%

Synonym(s):

(2S,2′S)-2,2′-Bipyrrolidine (2S,3S)-2,3-dihydroxybutanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2 · C4H6O6 · 3H2O
CAS Number:
Molecular Weight:
344.36
Beilstein:
5387052
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (sum of enantiomers, GC)
99%

form

crystals

optical purity

ee: ≥99.5% (GC)

mp

117-118 °C (dec.) (lit.)

SMILES string

O.O.O.O[C@@H]([C@H](O)C(O)=O)C(O)=O.C1CNC(C1)C2CCCN2

InChI

1S/C8H16N2.C4H6O6.3H2O/c1-3-7(9-5-1)8-4-2-6-10-8;5-1(3(7)8)2(6)4(9)10;;;/h7-10H,1-6H2;1-2,5-6H,(H,7,8)(H,9,10);3*1H2/t7-,8-;1-,2-;;;/m00.../s1

InChI key

XWLGYXKNZXAYBR-KVNOGXSSSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service