Skip to Content
MilliporeSigma
All Photos(1)

Documents

688533

Sigma-Aldrich

(S)-2-[2-[Bis(2-tolyl)phosphino]phenyl]-4-tert-butyl-2-oxazoline

97%

Synonym(s):

(S)-2-[2-[Bis(2-tolyl)phosphino]phenyl]-4-tert-butyl-4,5-dihydro-oxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30NOP
CAS Number:
Molecular Weight:
415.51
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]22/D −69.0, c = 1 in chloroform

mp

134-138 °C

storage temp.

−20°C

SMILES string

Cc1ccccc1P(c2ccccc2C)c3ccccc3C4=N[C@H](CO4)C(C)(C)C

InChI

1S/C27H30NOP/c1-19-12-6-9-15-22(19)30(23-16-10-7-13-20(23)2)24-17-11-8-14-21(24)26-28-25(18-29-26)27(3,4)5/h6-17,25H,18H2,1-5H3/t25-/m1/s1

InChI key

PWDWTBOBIGNWSX-RUZDIDTESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Andreas Pfaltz has a longstanding interest in the design of chiral ligands for asymmetric catalysis. The semicorrins developed in his group served as the prototype for an important family of nitrogen ligands, the bisoxazolines, which have found widespread use in catalytic asymmetric synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service