Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

686654

Sigma-Aldrich

Polyester-32-hydroxyl-1-carboxyl bis-MPA dendron, generation 5

≥97%

Synonym(s):

Dendron-G5-Carboxyl-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C155H250O94
Molecular Weight:
3617.59
UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

Quality Level

Assay

≥97%

form

powder (lyophilized)

mol wt

generation 5

feature

focal point carboxylic acid

no. Surface Groups

32

storage temp.

2-8°C

General description

Soluble in a variety of polar solvents, such as DMF, methanol, and water.
calculated mol. wt. 3,618

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service