Skip to Content
MilliporeSigma
All Photos(1)

Documents

682381

Sigma-Aldrich

Grubbs Catalyst® M360

Synonym(s):

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene][3-(2-pyridinyl) propylidene]ruthenium(II), Grubbs Catalyst® C598, [1,3-Dimesityl-2-imidazolidinylidene]dichloro[3-(2-pyridinyl)propylidene]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H35Cl2N3Ru
CAS Number:
Molecular Weight:
597.58
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

mp

190.5-199.0 °C

Quality Level

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(N2CCN(C2[Ru](Cl)(Cl)=CCCc3ccccn3)c4c(C)cc(C)cc4C)c(C)c1

InChI

1S/C21H27N2.C8H9N.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-2-5-8-6-3-4-7-9-8;;;/h9-13H,7-8H2,1-6H3;1,3-4,6-7H,2,5H2;2*1H;/q;;;;+2/p-2

InChI key

LCRADYBTPYSTNK-UHFFFAOYSA-L

Application

Metathesis: Ruthenium-Based Metathesis Catalysts

  • Catalyst for olefin metathesis reactions
  • Latent initiator, but extremely active once initiated, similar to other 2nd generation catalysts.
  • Useful in ROMP applications where longer monomer/catalyst resin handling times are desired.
  • Potentially useful in non-ROMP applications as well.
  • Enabling metathesis catalyst.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at www.pmc.umicore.com.
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ung, T. et al.
Organometallics, 23, 5399-5399 (2004)
Benitez, D.; Goddard, W. A., III.
Journal of the American Chemical Society, 12218-12218 (2005)
Schrodi, Y.; Pederson, R.L.
Aldrichimica Acta, 40, 45-45 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service