Skip to Content
MilliporeSigma
All Photos(1)

Documents

680184

Sigma-Aldrich

(S)-3,3′-Bis(triphenylsilyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate

96%

Synonym(s):

(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho-[2,1-d1′,2′f]-[1,3,2]-dioxaphosphepin 4-oxide, MacMillan TiPSY catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H41O4PSi2
CAS Number:
Molecular Weight:
865.07
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]22/D +210°, c = 1 in chloroform

mp

188-193 °C

SMILES string

OP1(=O)Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)[Si](c6ccccc6)(c7ccccc7)c8ccccc8)[Si](c9ccccc9)(c%10ccccc%10)c%11ccccc%11

InChI

1S/C56H41O4PSi2/c57-61(58)59-55-51(62(43-25-7-1-8-26-43,44-27-9-2-10-28-44)45-29-11-3-12-30-45)39-41-23-19-21-37-49(41)53(55)54-50-38-22-20-24-42(50)40-52(56(54)60-61)63(46-31-13-4-14-32-46,47-33-15-5-16-34-47)48-35-17-6-18-36-48/h1-40H,(H,57,58)

InChI key

BDQOCXQVIFQJRK-UHFFFAOYSA-N

Application

(S)-3,3′-Bis(triphenylsilyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate can be used as a catalyst:
  • In the synthesis of 1,3-dioxolochroman skeletons by reacting 3-methyl-2-vinylindoles with ortho-quinone methides.
  • In the exo-selective cyclization of mupirocin methyl ester.
  • In the kinetic resolution of cyclic aliphatic syn-1,3-diols, which are used as valuable building blocks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereo-and Enantioselective Construction of Biologically Important Chiral 1, 3-Dioxolochroman Frameworks via Catalytic Asymmetric [4+ 2] Cycloaddition
Tu M, et al.
The Journal of Organic Chemistry, 85(8), 5403-5415 (2020)
Parallel Kinetic Resolution of Unsymmetrical Acyclic Aliphatic syn-1, 3-Diols
Yang H and Zheng W
Organic Letters, 21(13), 5197-5200 (2019)
Experimental and Computational Studies on Regiodivergent Chiral Phosphoric Acid Catalyzed Cycloisomerization of Mupirocin Methyl Ester
Wang S, et al.
Chemistry?A European Journal , 26(8), 4583-4591 (2020)

Articles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service