Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

679453

Sigma-Aldrich

2-(Bromomethyl)phenylboronic acid

Synonym(s):

α-Bromo-o-tolueneboronic acid, o-Boronobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BBrO2
CAS Number:
Molecular Weight:
214.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

158-162 °C

Quality Level

functional group

bromo

storage temp.

2-8°C

SMILES string

OB(O)c1ccccc1CBr

InChI

1S/C7H8BBrO2/c9-5-6-3-1-2-4-7(6)8(10)11/h1-4,10-11H,5H2

InChI key

MYVJCOQGXCONPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride.

Application

2-(Bromomethyl)phenylboronic acid can be used as a reactant to synthesize:
  • Boronic acid-functionalized benzyl viologen (o-BBV) from 4,4′-bipyridine. o-BBV finds its application as a chemosensor to sense glucose in aqueous water.
  • 2-(azidomethyl)phenylboronic acid, which is further employed in the preparation of isoquinoline derivatives.
  • Aryl boronic acid derivatives as fluorescent probe for hydrogen peroxide detection.

Reactant involved in:
  • Studies of carbon-boron bond cleavage of phenylboronate-pendant cyclen
  • Development of a sensory system to detect glucose or other monosaccharides and hydroxycarboxylates
  • Synthesis of boronated triaryl and tetraaryl phosphonium salts used in cytotoxicity studies
  • Colorimetry and fluorometry mercury determination with chemosensors composed of rhodamine and boronic acid groups
  • Synthesis of imidazolium-containing boronic acids used as fluoride ion sensors
  • Reactions with adenine for molecules with antiinflammatory antitumor activities 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A highly sensitive water-soluble system to sense glucose in aqueous solution
Feng L, et al.
Organic & Biomolecular Chemistry, 9(8), 2938-2942 (2011)
2-(Azidomethyl) phenylboronic acid in the synthesis of isoquinoline derivatives
Ganina OG, et al.
Russian Chemical Bulletin, 54(7), 1606-1611 (2005)
A simple boronic acid-based fluorescent probe for selective detection of hydrogen peroxide in solutions and living cells
Han J, et al.
Bioorganic Chemistry, 81(7), 362-366 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service