Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

675229

Sigma-Aldrich

5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O2
CAS Number:
Molecular Weight:
354.53
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

164-169 °C

SMILES string

Cc1cc(c(O)c(c1C)-c2c(C)c(C)cc(c2O)C(C)(C)C)C(C)(C)C

InChI

1S/C24H34O2/c1-13-11-17(23(5,6)7)21(25)19(15(13)3)20-16(4)14(2)12-18(22(20)26)24(8,9)10/h11-12,25-26H,1-10H3

InChI key

NMVVBVMYPLMIOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Cocatalyst in titanium-catalyzed intermolecular hydroamination

Reactant in preparation of:
  • Sulfonate phenol ligands via sulfonylation
  • Catalysts for asymmetric olefin hydroamination/cyclization reactions
  • Single site catalyst precursors for catalysis of ring-opening reactions
  • Biphen-Mo complex used for catalysis of enantioselective ring-closing metathesis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service