Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

666491

Sigma-Aldrich

2,4-Dimethoxy-5-pyrimidinylboronic acid

90%

Synonym(s):

(2,4-Dimethoxypyrimidin-5-yl)boronic acid, (2,6-Dimethoxypyrimidin-5-yl)boronic acid, 2,4-Dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxy-5-Pyrimidineboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2(C4HN2)B(OH)2
CAS Number:
Molecular Weight:
183.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

impurities

5-bromo-2,4-dimethoxypyrimidine

mp

113-117 °C

storage temp.

2-8°C

SMILES string

COc1ncc(B(O)O)c(OC)n1

InChI

1S/C6H9BN2O4/c1-12-5-4(7(10)11)3-8-6(9-5)13-2/h3,10-11H,1-2H3

InChI key

LKGKUACPLXCVOF-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service