Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662739

Sigma-Aldrich

4-Bromo-2-methylpyridine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.556

density

1.450 g/mL at 25 °C

functional group

bromo

SMILES string

Cc1cc(Br)ccn1

InChI

1S/C6H6BrN/c1-5-4-6(7)2-3-8-5/h2-4H,1H3

InChI key

JFBMFWHEXBLFCR-UHFFFAOYSA-N

Application

Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.0 °F - closed cup

Flash Point(C)

78.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 907-907 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service