Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

656291

Sigma-Aldrich

4,5-Dibromo-2-furoic acid

97%

Synonym(s):

2,3-Dibromofuran-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2Br2O3
CAS Number:
Molecular Weight:
269.88
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

169-173 °C

functional group

bromo
carboxylic acid

SMILES string

OC(=O)c1cc(Br)c(Br)o1

InChI

1S/C5H2Br2O3/c6-2-1-3(5(8)9)10-4(2)7/h1H,(H,8,9)

InChI key

BHUVICYZDBUMIU-UHFFFAOYSA-N

Application

Used in a study of the effect of a furanyl halo-substituent on the intramolecular Diels-Alder reaction between the furan ring system and a pendant allylamide. The yield of the cycloaddition is improved with a C-5 bromide vs -chloro or -unsubstituted furan.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Albert Padwa et al.
The Journal of organic chemistry, 71(15), 5432-5439 (2006-07-15)
Intramolecular Diels-Alder reactions involving a series of N-alkenyl-substituted furanyl amides were investigated. Stable functionalized oxanorbornenes were formed in high yield upon heating at 80-110 degrees C. The cycloaddition reactions include several bromo-substituted furanyl amides, and these systems were found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service