Skip to Content
MilliporeSigma
All Photos(1)

Documents

650331

Sigma-Aldrich

Cyclopentylacetylene

90%

Synonym(s):

1-Ethynylcyclopentane, Ethynylcyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10
CAS Number:
Molecular Weight:
94.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

refractive index

n20/D 1.432 (lit.)

density

0.812 g/mL at 25 °C (lit.)

SMILES string

C#CC1CCCC1

InChI

1S/C7H10/c1-2-7-5-3-4-6-7/h1,7H,3-6H2

InChI key

TXVJSWLZYQMWPC-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

34.0 °F

Flash Point(C)

1.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service