Skip to Content
MilliporeSigma
All Photos(1)

Documents

649805

Sigma-Aldrich

PEDOT, block PEG

solution, 1 wt. % dispersion in nitromethane, contains perchlorate as dopant

Synonym(s):

Aedotron C-NM, PEDOT-block-PEG, PEDOT-b-PEG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene)-block-poly(ethylene glycol) solution, 1 wt % dispersion in nitromethane, contains perchlorate as dopant

contains

perchlorate as dopant

Quality Level

concentration

1 wt % dispersion in nitromethane

resistance

10,000-100,000 Ω/sq (spin cast thin films: typically 1-3 layers spun at 1,000 rpm)

work function

4.33 eV

description

40 nm (RMS roughness spin cast thin films)

particle size

600-1000 nm (in suspension)

conductivity

0.1-5.0 S/cm (bulk)

density

1.127 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Aedotron polymers are solvent-dispersable forms of PEDOT. These are copolymers of doped PEDOT and a flexible, soluble polymer such as poly(ethylene glycol) (PEG). These colloidal dispersions are stabilized by the highly solvated PEG chains which sterically limit the aggregation of the PEDOT blocks. Aedotron polymers are neither acidic nor corrosive, it easily disperses in polar aprotic solvents. Perchlorate-doped copolymers typically have a higher conductivity with thin films that are more transparent.
Please follow this link for Aedotron/Oligotron selection guide table.

Application

Nitromethane dispersions can be used to cast thin films on glass, ITO, silicon wafers and many polymeric substrates. Compatibility of the polymeric substrate with nitromethane should be tested in each case. Coat the substrate with a "pool" of the nitromethane dispersion. Thin transparent films are typically spun at 1000 RPM or higher speeds, but more conducting films can be obtained at lower speeds. A single layer spun at 1000 RPM is typically 75 nm thick.
Conducting polymer. Useful as anti-static layers, hole-transport layers in OLED′s and various organic electronics applications (flexible circuits, capacitors, batteries).

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Aedotron is a trademark of TDA Research, Inc.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

In the field of organic printable electronics, such as OLEDs and organic photovoltaics (OPVs), improved organic conducting and semiconducting materials are needed. The progress in two fields is reviewed in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service