Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

645532

Sigma-Aldrich

4-Cyanoindole

97%

Synonym(s):

Indole-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

117-121 °C (lit.)

functional group

nitrile

SMILES string

N#Cc1cccc2[nH]ccc12

InChI

1S/C9H6N2/c10-6-7-2-1-3-9-8(7)4-5-11-9/h1-5,11H

InChI key

CEUFGDDOMXCXFW-UHFFFAOYSA-N

Application

Reactant for preparation of tryptophan dioxygenase inhibitors as potential anticancer immunomodulators
Reactant for stereoselective preparation of γ-lactones via prolinol silyl ether catalyzed Friedel-Crafts alkylation reaction
Reactant for iridium-catalyzed borylation reactions
Reactant for parallel synthesis of N-benzylisatin oximes as JNK3 MAP kinase inhibitors
Reactant for preparation of N-aryl indoles
Reactant for preparation of conformationally restricted indole cyclopropylmethylamines as selective serotonin reuptake inhibitors

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service