Skip to Content
MilliporeSigma
All Photos(1)

Documents

639370

Sigma-Aldrich

4-Methoxy-2-methylphenylboronic acid

≥95%

Synonym(s):

2-Methyl-4-methoxybenzeneboronic acid, 2-Methyl-4-methoxyphenylboronic acid, 4-Methoxy-2-methylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)B(OH)2
CAS Number:
Molecular Weight:
165.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

169-174 °C (lit.)

SMILES string

COc1ccc(B(O)O)c(C)c1

InChI

1S/C8H11BO3/c1-6-5-7(12-2)3-4-8(6)9(10)11/h3-5,10-11H,1-2H3

InChI key

AMSQNQJCBXQYEX-UHFFFAOYSA-N

Application

Reactant for preparation of biologically active molecules:
  • Preparation of hydroxyphenylnaphthols as 17ß-hydroxysteroid dehydrogenase Type 2 inhibitors

Reactant for:
  • Preparation of axially-chiral biarylphosphonates by asymmetric Suzuki coupling catalyzed by palladium complexes with helically-chiral polyquinoxaline phosphine and polyphosphine copolymers
  • Preparation of arylpyrimidines and their use in regioselective acetoxylation
  • Preparation of N-arylated azoles via copper fluorapatite-catalyzed arylation

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service