Skip to Content
MilliporeSigma

Skip To

634530

5-Bromo-1,2,3-trimethoxybenzene

97%

Synonym(s):

1-Bromo-3,4,5-trimethoxybenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H11BrO3
CAS Number:
Molecular Weight:
247.09
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
220-223-9
MDL number:
Assay:
97%
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

solid

mp

78-82 °C (lit.)

functional group

bromo

SMILES string

COc1cc(Br)cc(OC)c1OC

InChI

1S/C9H11BrO3/c1-11-7-4-6(10)5-8(12-2)9(7)13-3/h4-5H,1-3H3

InChI key

XAOOZMATJDXDQJ-UHFFFAOYSA-N

Application

5-Bromo-1,2,3-trimethoxybenzene can be used to synthesize the following:
  • analogs of HA14-1, which shows promising anticancer properties
  • N,N′-diarylated indolo[3,2-b]carbazole derivatives, which can find applications in electrophotography
  • symmetric 3,3′,4,4′,5,5′-hexamethoxydiphenylacetylene via Stille-type coupling with bis-(tributylstannyl)acetylene

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
330841759341708453
description

97%

description

99%

description

97%

description

97%

assay

97%

assay

99%

assay

97%

assay

97%

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

mp

78-82 °C (lit.)

mp

-

mp

-

mp

75-80 °C

form

solid

form

liquid

form

liquid

form

solid

functional group

bromo

functional group

bromo, fluoro

functional group

bromo, fluoro

functional group

bromo, ketone


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters