Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

633054

Sigma-Aldrich

2-Acetyl-3-bromothiophene

97%

Synonym(s):

1-(3-Bromo-2-thienyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrOS
CAS Number:
Molecular Weight:
205.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

refractive index

n20/D 1.612

mp

28-35 °C

functional group

bromo
ketone

SMILES string

CC(=O)c1sccc1Br

InChI

1S/C6H5BrOS/c1-4(8)6-5(7)2-3-9-6/h2-3H,1H3

InChI key

ZUXPELAHJQSZTE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khyati P Thakor et al.
Luminescence : the journal of biological and chemical luminescence, 34(1), 113-124 (2019-01-04)
Novel palladium(II) complexes (7a-7e) of substituted quinoline derivatives were synthesized. The complexes were characterized using various techniques such as thermogravimetric analysis (TGA), elemental analysis, conductance measurement, mass, absorption, infra-red (IR), 1 H NMR, 13 C NMR and energy-dispersive X-ray spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service