Skip to Content
MilliporeSigma

632007

Isobutyric-d7 acid

98 atom % D, 99% (CP)

Synonym(s):

2-Methylpropionic-d7 acid

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CD3CD(CD3)CO2H
CAS Number:
Molecular Weight:
95.15
UNSPSC Code:
51113400
PubChem Substance ID:
MDL number:
Isotopic purity:
98 atom % D
Assay:
99% (CP)
Mass shift:
M+7


isotopic purity

98 atom % D

assay

99% (CP)

bp

153-154 °C (lit.)

mp

-47 °C (lit.)

density

1.024 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(C(O)=O)C([2H])([2H])[2H]

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i1D3,2D3,3D

InChI key

KQNPFQTWMSNSAP-YYWVXINBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.



Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

131.0 °F - closed cup

Flash Point (°C)

55 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library